Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ser-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029164.3 | NP_001024335.1 | 5 - 6 | 535 | 103 | |
NM_001029165.5 | NP_001024336.1 | 5 - 6 | 1011 | 103 | ||
NM_001029168.4 | NP_001024339.1 | 5 - 6 | 535 | 103 | ||
NM_001029169.5 | NP_001024340.1 | 4 - 5 | 620 | 103 | ||
NM_001270331.4 | NP_001257260.1 | 5 - 6 | 535 | 103 | ||
GenBank mRNA | GQ217520.1 | - | 4 - 5 | 502 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |