Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029782.5 | NP_001024953.1 | 5 - 6 | 509 | 64 | |
NM_001383682.1 | 5 - 6 | 514 | 64 | |||
GenBank mRNA | AY349017.1 | - | 5 - 6 | 531 | 64 | |
AY349018.1 | - | 2 - 3 | 171 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |