Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: svh-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270332.3 | NP_001257261.1 | 7 - 8 | 1381 | 93 | |
NM_001383683.1 | 8 - 9 | 1628 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |