Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029570.6 | NP_001024741.1 | 8 - 9 | 1135 | 65 | |
NM_001383685.1 | 9 - 10 | 1187 | 65 | |||
NR_002425.1 | 8 - 9 | 1031 | 65 | |||
GenBank mRNA | AB032748.1 | - | 8 - 9 | 1133 | 65 | |
AB032749.1 | - | 8 - 9 | 1133 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |