Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olrn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029163.7 | NP_001024334.1 | 5 - 6 | 1458 | 64 | |
NM_001373597.2 | NP_001359691.1 | 5 - 6 | 1444 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |