Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gas-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078168.6 | NP_510569.1 | 5 - 6 | 1322 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |