Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipi-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078172.9 | NP_510573.3 | 2 - 3 | 784 | 113 | |
GenBank mRNA | EU043523.1 | - | 2 - 3 | 781 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |