Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373609.3 | NP_001360585.1 | 14 | 3438 | 106 | |
NM_078249.7 | NP_510650.2 | 13 - 14 | 3426 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |