Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373610.3 | NP_001360030.1 | 18 - 19 | 2354 | 114 |
Gene Symbol: F38E9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078272.7 | NP_510673.1 | 8 - 9 | 1109 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |