Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373610.3 | NP_001360030.1 | 20 - 21 | 2564 | 68 |
Gene Symbol: F38E9.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078272.7 | NP_510673.1 | 10 - 11 | 1319 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |