Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078280.8 | NP_510681.4 | 23 - 24 | 5645 | 75 | |
GenBank mRNA | DQ360811.1 | - | 23 - 24 | 4929 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |