Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hda-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270349.3 | NP_001257278.1 | 4 - 5 | 876 | 87 | |
NM_001270350.5 | NP_001257279.1 | 4 - 5 | 878 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |