Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078311.8 | NP_510712.2 | 8 - 9 | 1474 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |