Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cas-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373613.3 | NP_001360035.1 | 2 - 3 | 3139 | 86 | |
NM_078312.6 | NP_510713.1 | 3 - 4 | 333 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |