Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078421.6 | NP_510822.1 | 9 - 10 | 1677 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |