Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047837.4 | NP_001041302.1 | 18 - 19 | 3401 | 74 | |
NM_001047838.4 | NP_001041303.1 | 17 - 18 | 3233 | 74 | ||
GenBank mRNA | AJ512332.1 | - | 18 - 19 | 3344 | 74 | |
AJ811573.1 | - | 18 - 19 | 3372 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |