Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047837.4 | NP_001041302.1 | 2 - 3 | 218 | 87 | |
NM_001047838.4 | NP_001041303.1 | 1 - 2 | 50 | 87 | ||
GenBank mRNA | AJ512332.1 | - | 2 - 3 | 161 | 87 | |
AJ811573.1 | - | 2 - 3 | 189 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |