Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078456.5 | NP_510857.2 | 6 - 7 | 1250 | 111 | |
GenBank mRNA | AF364401.1 | - | 6 - 7 | 1254 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |