Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wwp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306342.3 | NP_001293271.1 | 2 - 3 | 434 | 112 | |
NM_001306343.3 | NP_001293272.1 | 2 - 3 | 428 | 112 | ||
NM_170794.7 | NP_740775.1 | 6 - 7 | 1311 | 112 | ||
NM_171831.4 | NP_740776.1 | 6 - 7 | 1305 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |