Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306422.3 | NP_001293351.1 | 6 - 7 | 1316 | 73 | |
NM_170819.6 | NP_740802.1 | 8 - 9 | 1713 | 73 | ||
NR_131352.1 | 8 - 9 | 1552 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |