Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377804.1 | NP_001364770.1 | 5 - 6 | 985 | 104 | |
NM_001381436.2 | NP_001367838.1 | 5 - 6 | 973 | 104 | ||
NM_170830.6 | NP_740814.2 | 5 - 6 | 972 | 104 | ||
NR_131363.1 | 5 - 6 | 943 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |