Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373813.1 | NP_001360094.1 | 2 - 3 | 274 | 119 | |
NM_058775.7 | NP_491176.1 | 2 - 3 | 280 | 119 | ||
NM_170843.5 | NP_740829.1 | 2 - 3 | 264 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |