Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noah-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263093.4 | NP_001250022.1 | 1 - 2 | 123 | 100 | |
NM_170870.8 | NP_740858.2 | 1 - 2 | 119 | 100 | ||
NM_171838.7 | NP_740859.3 | 1 - 2 | 119 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |