Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380502.1 | NP_001367496.1 | 3 - 4 | 387 | 116 | |
NM_170878.5 | NP_740867.1 | 3 - 4 | 374 | 116 | ||
NM_170879.8 | NP_740868.1 | 3 - 4 | 369 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |