Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025804.5 | NP_001020975.1 | 1 - 2 | 247 | 75 | |
GenBank mRNA | AF332203.1 | - | 1 - 2 | 158 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |