Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025804.5 | NP_001020975.1 | 2 - 3 | 634 | 112 | |
NM_001037882.5 | NP_001032971.1 | 1 - 2 | 390 | 112 | ||
NM_001380645.1 | NP_001367012.1 | 1 - 2 | 403 | 112 | ||
GenBank mRNA | AF332203.1 | - | 2 - 3 | 545 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |