Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025804.5 | NP_001020975.1 | 8 - 9 | 1857 | 87 | |
NM_001025806.6 | NP_001020977.1 | 7 - 8 | 1477 | 87 | ||
NM_001037880.5 | NP_001032969.1 | 6 - 7 | 1232 | 87 | ||
NM_001037882.5 | NP_001032971.1 | 7 - 8 | 1619 | 87 | ||
NM_001380645.1 | NP_001367012.1 | 7 - 8 | 1626 | 87 | ||
NM_001380646.1 | NP_001367128.1 | 7 - 8 | 1615 | 87 | ||
GenBank mRNA | AF332203.1 | - | 8 - 9 | 1768 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Endogenous Controls Detect Genomic DNA Caenorhabditis elegans Probe spans exons |