Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026412.4 | NP_001021583.1 | 3 - 4 | 460 | 150 | |
GenBank mRNA | AB040992.1 | - | 3 - 4 | 465 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |