Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170938.6 | NP_740934.2 | 3 - 4 | 458 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |