Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170941.7 | NP_740937.1 | 4 - 5 | 1103 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |