Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: catp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129056.5 | NP_001122528.1 | 12 - 13 | 2692 | 69 | |
NM_001129057.3 | NP_001122529.1 | 12 - 13 | 2691 | 69 | ||
GenBank mRNA | AM419897.1 | - | 12 - 13 | 2681 | 69 | |
AM419898.1 | - | 12 - 13 | 2681 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |