Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381273.1 | NP_001367025.1 | 1 - 2 | 108 | 87 | |
NM_170962.5 | NP_740958.1 | 1 - 2 | 246 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |