Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381274.1 | NP_001367022.1 | 1 - 2 | 50 | 68 | |
NM_170966.3 | NP_740962.1 | 1 - 2 | 379 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |