Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047328.5 | NP_001040793.1 | 3 - 4 | 394 | 67 | |
NM_001047329.4 | NP_001040794.1 | 3 - 4 | 238 | 67 | ||
NM_001381394.1 | NP_001367817.1 | 4 - 5 | 443 | 67 | ||
NM_001381395.2 | NP_001367816.1 | 4 - 5 | 447 | 67 | ||
GenBank mRNA | EU180219.1 | - | 4 - 5 | 427 | 67 |
Gene Symbol: coa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383820.1 | 6 - 7 | 1028 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |