Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171004.7 | NP_741003.1 | 6 - 7 | 877 | 120 | |
NR_136362.1 | 6 - 7 | 877 | 120 | |||
NR_136363.1 | 5 - 6 | 1152 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |