Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171004.7 | NP_741003.1 | 7 - 8 | 1026 | 141 | |
NR_136363.1 | 6 - 7 | 1301 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |