Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381440.1 | NP_001367789.1 | 1 - 2 | 261 | 104 | |
NM_062780.6 | NP_495181.2 | 3 - 4 | 432 | 104 | ||
NM_062781.5 | NP_495182.2 | 2 - 3 | 356 | 104 | ||
NR_151564.1 | 2 - 3 | 325 | 104 | |||
GenBank mRNA | DQ178632.1 | - | 261 | 104 | ||
DQ178633.1 | - | 432 | 104 | |||
DQ178634.1 | - | 372 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |