Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356848.3 | NP_001343765.1 | 3 | 177 | 70 | |
NM_001356849.3 | NP_001343766.1 | 3 | 177 | 70 | ||
NM_001356850.3 | NP_001343767.1 | 3 | 180 | 70 | ||
NM_001381441.2 | NP_001367307.1 | 2 - 3 | 145 | 70 | ||
NM_062785.5 | NP_495186.3 | 3 - 4 | 205 | 70 | ||
NM_171007.5 | NP_741006.3 | 3 - 4 | 205 | 70 | ||
NM_171850.7 | NP_741007.2 | 3 - 4 | 224 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |