Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356848.3 | NP_001343765.1 | 4 | 376 | 133 | |
NM_001356849.3 | NP_001343766.1 | 4 | 376 | 133 | ||
NM_001356850.3 | NP_001343767.1 | 4 | 379 | 133 | ||
NM_001356852.3 | NP_001343768.1 | 3 - 4 | 324 | 133 | ||
NM_001381441.2 | NP_001367307.1 | 3 - 4 | 344 | 133 | ||
NM_001381442.1 | NP_001368664.1 | 1 - 2 | 205 | 133 | ||
NM_001383853.1 | 3 - 4 | 340 | 133 | |||
NM_062785.5 | NP_495186.3 | 4 - 5 | 404 | 133 | ||
NM_171007.5 | NP_741006.3 | 4 - 5 | 404 | 133 | ||
NM_171850.7 | NP_741007.2 | 4 - 5 | 423 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |