Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351913.2 | NP_001338820.1 | 3 - 4 | 480 | 77 | |
NM_001351914.2 | NP_001338821.1 | 3 - 4 | 589 | 77 | ||
NM_001381477.2 | NP_001367332.1 | 3 - 4 | 449 | 77 | ||
NM_001381478.2 | NP_001368653.1 | 2 - 3 | 416 | 77 | ||
NM_001381479.1 | NP_001368654.1 | 2 - 3 | 411 | 77 | ||
NM_001381480.1 | NP_001368655.1 | 2 - 3 | 416 | 77 | ||
NM_001381481.1 | NP_001368656.1 | 2 - 3 | 344 | 77 | ||
NM_171852.2 | NP_741018.1 | 3 - 4 | 475 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |