Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171019.3 | NP_741021.1 | 11 - 12 | 1675 | 69 | |
NM_171020.5 | NP_741022.1 | 11 - 12 | 1665 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |