Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026736.5 | NP_001021907.1 | 7 - 8 | 2276 | 127 | |
NM_001026737.5 | NP_001021908.1 | 7 - 8 | 2308 | 127 | ||
NM_001383915.1 | 7 - 8 | 2271 | 127 | |||
GenBank mRNA | AJ132946.1 | - | 7 - 8 | 2275 | 127 | |
AJ132947.1 | - | 6 - 7 | 2196 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |