Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171035.6 | NP_741041.1 | 7 - 8 | 861 | 81 | |
NM_171036.6 | NP_741042.1 | 2 - 3 | 434 | 81 | ||
GenBank mRNA | LC026472.1 | - | 6 - 7 | 805 | 81 | |
LC026473.1 | - | 2 - 3 | 319 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |