Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 11 - 12 | 1547 | 97 | |
NM_001026999.8 | NP_001022170.1 | 11 - 12 | 1547 | 97 | ||
NM_001027000.6 | NP_001022171.1 | 8 - 9 | 1342 | 97 | ||
NM_001027001.5 | NP_001022172.1 | 3 - 4 | 287 | 97 | ||
NM_001267342.3 | NP_001254271.1 | 11 - 12 | 1547 | 97 | ||
NM_001267343.3 | NP_001254272.1 | 11 - 12 | 1547 | 97 | ||
NM_001267344.3 | NP_001254273.1 | 4 - 5 | 623 | 97 | ||
NM_171039.3 | NP_741045.1 | 11 - 12 | 1588 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |