Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 21 - 22 | 4421 | 101 | |
NM_001026999.8 | NP_001022170.1 | 22 - 23 | 4736 | 101 | ||
NM_001027000.6 | NP_001022171.1 | 19 - 20 | 4531 | 101 | ||
NM_001027001.5 | NP_001022172.1 | 14 - 15 | 3476 | 101 | ||
NM_001267342.3 | NP_001254271.1 | 20 - 21 | 4400 | 101 | ||
NM_001267343.3 | NP_001254272.1 | 21 - 22 | 4613 | 101 | ||
NM_001267344.3 | NP_001254273.1 | 15 - 16 | 3812 | 101 | ||
NM_171039.3 | NP_741045.1 | 21 - 22 | 4564 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |