Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 5 - 6 | 418 | 85 | |
NM_001026999.8 | NP_001022170.1 | 5 - 6 | 418 | 85 | ||
NM_001027000.6 | NP_001022171.1 | 2 - 3 | 213 | 85 | ||
NM_001267342.3 | NP_001254271.1 | 5 - 6 | 418 | 85 | ||
NM_001267343.3 | NP_001254272.1 | 5 - 6 | 418 | 85 | ||
NM_171039.3 | NP_741045.1 | 5 - 6 | 459 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |