Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pll-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171059.6 | NP_741067.1 | 18 - 19 | 2912 | 63 | |
NM_171060.5 | NP_741068.1 | 18 - 19 | 2940 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |