Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zer-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171078.7 | NP_741089.2 | 5 - 6 | 2536 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Caenorhabditis elegans Probe spans exons |