Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065271.6 | NP_497672.2 | 8 - 9 | 1624 | 88 | |
NM_171096.6 | NP_741109.1 | 9 - 10 | 1748 | 88 | ||
NM_171097.5 | NP_741110.1 | 8 - 9 | 1636 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |