Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065331.4 | NP_497732.1 | 15 - 16 | 2757 | 89 | |
NM_065332.4 | NP_497733.1 | 8 - 9 | 1568 | 89 | ||
NM_171099.5 | NP_741112.1 | 8 - 9 | 1401 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |